Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRED1 Antibody (M23-P2G3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250308V
Description
SPRED1 Monoclonal specifically detects SPRED1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SPRED1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPRED1 | |
Peptide Sequence YVPLRMCHR (amino acids 420 - 428) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
M23-P2G3 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation | |
Mouse | |
Protein A purified | |
RUO | |
161742 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction