Learn More
Invitrogen™ SPRED1 Polyclonal Antibody

Description
Immunogen sequence: GQPGLDIQSR SMEYVQRQIS KECGSLKSQN RVPLKSIRHV SFQDEDEIVR INPRDILIRR YADYRHPDMW KNDLERDDAD SSI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Specifications
Specifications
| Antigen | SPRED1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SPRED1 |
| Gene Accession No. | Q7Z699 |
| Gene Alias | 5730461F13Rik; AL024345; AW047647; hSpred1; NFLS; PPP1R147; protein phosphatase 1, regulatory subunit 147; SPRED1; spred-1; sprouty protein with EVH-1 domain 1, related sequence; sprouty related EVH1 domain containing 1; sprouty related, EVH1 domain containing 1; sprouty-related protein 1 with EVH-1 domain; sprouty-related, EVH1 domain containing 1; sprouty-related, EVH1 domain-containing protein 1; suppressor of Ras/MAPK activation |
| Gene Symbols | SPRED1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.