Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598571
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598571 Supplier Invitrogen™ Supplier No. PA598571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRED1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The Ras-MAP kinase pathway is essential for the differentiation of neuronal cells and myocytes; it is inhibited by Spred1, a member of the Sprouty family of proteins. Spred1 acts by suppressing the phosphorylation and activation of Raf. The Spred proteins have also been implicated in the negative feedback regulation of FGF signaling in embryogenesis and angiogenesis. Further studies have shown that expression levels of Spred1 and Spred2 proteins are inversely correlated with the incidence of tumor invasion and metastasis in human hepatocellular carcinoma (HHC), suggesting that these proteins could be useful as prognostic factors and therapeutic targets in HCC. Defects in this gene are a cause of neurofibromatosis type 1-like syndrome (NFLS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRED1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPRED1
Gene Accession No. Q7Z699
Gene Alias 5730461F13Rik; AL024345; AW047647; hSpred1; NFLS; PPP1R147; protein phosphatase 1, regulatory subunit 147; SPRED1; spred-1; sprouty protein with EVH-1 domain 1, related sequence; sprouty related EVH1 domain containing 1; sprouty related, EVH1 domain containing 1; sprouty-related protein 1 with EVH-1 domain; sprouty-related, EVH1 domain containing 1; sprouty-related, EVH1 domain-containing protein 1; suppressor of Ras/MAPK activation
Gene Symbols SPRED1
Host Species Rabbit
Immunogen Recombinant Human Sprouty-related, EVH1 domain-containing protein 1 protein (123-311AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161742
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.