Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPRED2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531038
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (88%), Rat (88%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SPRED2 is a member of the Sprouty /SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (Nonami et al., 2004 ).
Specifications
| SPRED2 | |
| Polyclonal | |
| Unconjugated | |
| Spred2 | |
| C79158; Spred2; Spred-2; sprouty protein with EVH-1 domain 2, related sequence; sprouty related EVH1 domain containing 2; sprouty related, EVH1 domain containing 2; sprouty-related, EVH1 domain containing 2; sprouty-related, EVH1 domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.61 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z698 | |
| Spred2 | |
| Recombinant protein encompassing a sequence within the center region of human SPRED2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction