Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRED2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584640 Supplier Invitrogen™ Supplier No. PA584640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGSTTSSSTI HNEAELGDDD VFTTATDSSS NSSQKREQPT RTISSPTSCE HRRIYTLGHL HDSYPTDHYH LDQPMP Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 84%.

SPRED2 is a member of the Sprouty /SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (Nonami et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRED2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Spred2
Gene Accession No. Q7Z698
Gene Alias C79158; Spred2; Spred-2; sprouty protein with EVH-1 domain 2, related sequence; sprouty related EVH1 domain containing 2; sprouty related, EVH1 domain containing 2; sprouty-related, EVH1 domain containing 2; sprouty-related, EVH1 domain-containing protein 2
Gene Symbols Spred2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPRED2. Recombinant protein control fragment (Product #RP-104686).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.