Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986087
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6087 Supplier Bioss Supplier No. BS4566R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRN has PrP(C)-like neuroprotective activity. SPRN may act as a modulator for the biological actions of normal and abnormal PrP.

Specifications

Antigen SPRN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPRN
Gene Accession No. Q5BIV9
Gene Alias bA108K14.1; C630041J07; Gm169; hypothetical protein BC004409; Protein shadoo; SHADOO; Shadow of prion protein; shadow of prion protein homolog (zebrafish); SHO; SPRN
Gene Symbols SPRN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Shadow (CT).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 503542
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.