Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRR1B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526062

Catalog No. PIPA526062


Only null left
Add to Cart

Description

Description

SPRR1B Polyclonal Antibody for Western Blot, IHC (P)

SPRR1B is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane. Can function as both amine donor and acceptor in transglutaminase-mediated cross-linkage.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPRR1B
Polyclonal
Unconjugated
SPRR1B
14.9 kDa pancornulin; CORNIFIN; cornifin B; cornifin-B; GADD33; small proline rich protein 1B; small proline-rich protein 1B; Small proline-rich protein IB; SPR1 B; SPR1B; SPR-IB; SPRR1; SPRR1B
Rabbit
Antigen affinity chromatography, Protein A
RUO
6699
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P22528
SPRR1B
KLH conjugated synthetic peptide between 63-89 amino acids from the C-terminal region of human SPRR1B.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.