Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRR1B Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727595 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7595 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7595 Supplier Proteintech Group Inc Supplier No. 119591AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRR1B is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane. Can function as both amine donor and acceptor in transglutaminase-mediated cross-linkage.

Specifications

Antigen SPRR1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SPRR1B
Gene Accession No. P22528
Gene Alias 14.9 kDa pancornulin, CORNIFIN, Cornifin B, GADD33, Small proline rich protein IB, SPR IB, SPRR1, SPRR1B
Gene Symbols SPRR1B
Host Species Rabbit
Immunogen SPRR1B Fusion Protein Ag2563
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6699
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.