Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRR2A Antibody (OTI4B3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274347FR
Description
SPRR2A Monoclonal specifically detects SPRR2A in Human samples. It is validated for Western Blot.Specifications
SPRR2A | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
SPRR2A | |
Full length recombinant protein of human SPRR2A (NP_005979) produced in E.coli. | |
0.1 mL | |
6700.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4B3 | |
Western Blot | |
2-1, small proline-rich protein 2A, SPR-2A | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction