Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPRR2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107027
Description
Sequence homology: Guinea Pig: 79%; Human: 100%; Mouse: 83%; Rat: 83%.
Cross-linked envelope protein of keratinocytes. It is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane.
Specifications
| SPRR2A | |
| Polyclonal | |
| Unconjugated | |
| SPRR2A | |
| 1-Feb; 2-1; small proline rich protein 2A; small proline-rich protein 2A; SPR-2A; SPRR2A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6700 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P35326 | |
| SPRR2A | |
| Synthetic peptide located within the following region: CPEPCPPPKC PEPCPPPKCP QPCPPQQCQQ KYPPVTPSPP CQSKYPPKSK (aa 23-72). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction