Learn More
Invitrogen™ SPRR2F Monoclonal Antibody (5A9)

Description
Peptide Sequence: MSYQQQQCKQ PCQPPPVCPA PKCPEPCPPP KCPEPCPPSK CPQSCPPQQC QQKCPPVTPS PPCQPKCPPK SK.
Specifications
Specifications
| Antigen | SPRR2F |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5A9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SPRR2F |
| Gene Accession No. | Q96RM1 |
| Gene Alias | small proline rich protein 2F; small proline-rich protein 2F; SPR-2F; SPRR2F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.