Learn More
Invitrogen™ SPRR4 Monoclonal Antibody (1G5)

Description
Peptide Sequence: MSSQQQQRQQ QQCPPQRAQQ QQVKQPCQPP PVKCQETCAP KTKDPCAPQV KKQCPPKGTI IPAQQKCPSA QQASKSKQK.
Specifications
Specifications
| Antigen | SPRR4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G5 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SPRR4 |
| Gene Accession No. | Q96PI1 |
| Gene Alias | small proline rich protein 4; small proline-rich protein 4; SPRR4 |
| Gene Symbols | SPRR4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.