Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRTN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110424 Supplier Invitrogen™ Supplier No. PA5110424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDDDLMLALR LQEEWNLQEA ERDHAQESLS LVDASWELVD PTPDLQALFV QFNDQFFWGQ LEAVEVKWSV RMTLCAGICS YEGKGGMCSI RLSEPLLKLR PRKDLVETLL HEMIHAYLFV TNNDKDREGH GPEFCKHMHR INSLTGANIT VYHTFHDEVD EYRRHWWRCN GPCQHRPPYY GYVKRATNRE PSAHDYWWAE HQKTCGGTYI KIKEPENYSK KGKGKAKLGK EPVLAAENKG.

SPRTN is a regulator of UV-induced DNA damage response: acts as a 'reader' of ubiquitinated PCNA that enhances RAD18-mediated PCNA ubiquitination and translesion DNA synthesis. SPRTN is recruited to sites of UV damage and interacts with ubiquitinated PCNA and RAD18, the E3 ubiquitin ligase that monoubiquitinates PCNA. It facilitates chromatin association of RAD18 and is required for efficient PCNA monoubiquitination, promoting a feed-forward loop to enhance PCNA ubiquitination and translesion DNA synthesis. It is also required for localization of DNA polymerase eta (POLH) to sites of UV damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRTN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SPRTN
Gene Accession No. Q9H040
Gene Alias C1orf124; DDDL1880; dJ876B10.3; DNA damage protein targeting VCP; DNA damage-targeting VCP (p97) adaptor; DVC1; Gm505; PRO4323; protein with SprT-like domain at the N terminus; RGD1559496; spartan; sprT-like domain-containing protein Spartan; SprT-like N-terminal domain; Sprtn; UNQ1880/PRO4323; zinc finger RAD18 domain-containing protein
Gene Symbols SPRTN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SPRTN (NP_001010984.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83932
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.