Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565325 Supplier Invitrogen™ Supplier No. PA565325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTVVPRPGLKP APRPSTQHKH ERLHGLPEHR QPPRLQHSQV HSSARAPLSR SISTVSSGSR SSTRTSTSSS SSEQ Highest antigen sequence identity to the following orthologs - mouse 97%, rat 93%.

This gene encodes a protein belonging to the sprouty family. The encoded protein contains a carboxyl-terminal cysteine-rich domain essential for the inhibitory activity on receptor tyrosine kinase signaling proteins and is required for growth factor stimulated translocation of the protein to membrane ruffles. In primary dermal endothelial cells this gene is transiently upregulated in response to fibroblast growth factor two. This protein is indirectly involved in the non-cell autonomous inhibitory effect on fibroblast growth factor two signaling. The protein interacts with Cas-Br-M (murine) ectropic retroviral transforming sequence, and can function as a bimodal regulator of epidermal growth factor receptor/mitogen-activated protein kinase signaling. This protein may play a role in alveoli branching during lung development as shown by a similar mouse protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRY2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPRY2
Gene Accession No. O43597
Gene Alias hSPRY2; IGAN3; MGC23039; protein sprouty homolog 2; sprouty 2; sprouty homolog 2; sprouty homolog 2 (Drosophila); sprouty RTK signaling antagonist 2; sprouty2; SPRY2; spry-2
Gene Symbols SPRY2
Host Species Rabbit
Immunogen Recombinant Human SPRY2. Recombinant protein control fragment (Product #RP-106022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.