Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPRY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549294
Description
SPRY3 Polyclonal Antibody for Western Blot
May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.
Specifications
SPRY3 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O43610 | |
SPRY3 | |
KLH conjugated synthetic peptide between 261-288 amino acids from the C-terminal region of human SPRY3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SPRY3 | |
antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction