Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRY4 Antibody (SR1553), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | SPRY4 |
---|---|
Clone | SR1553 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
SPRY4 Monoclonal antibody specifically detects SPRY4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
SPRY4 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
protein sprouty homolog 4, sprouty homolog 4 (Drosophila), spry-4 | |
A synthesized peptide derived from human SPRY4 (Uniprot #: Q9C004) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1553 | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Growth and Development, Neuronal Cell Markers, Signal Transduction | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
81848 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title