Learn More
Invitrogen™ SPRY4 Polyclonal Antibody

Description
Immunogen sequence: SSDQRLLDHM APPPVADQAS PRAVRIQPKV VHCQPLDLKG PAVPPELDKH FLLCEACG Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
Specifications
Specifications
| Antigen | SPRY4 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SPRY4 |
| Gene Accession No. | Q9C004 |
| Gene Alias | A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4 |
| Gene Symbols | SPRY4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.