Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRYD3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177116
Description
SPRYD3 Polyclonal specifically detects SPRYD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SPRYD3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
FLJ14800, SPRY domain containing 3, SPRY domain-containing protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
84926 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin | |
NP_116229 | |
SPRYD3 | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of human SPRYD3. The immunogen is located within amino acids 260 - 310 of SPRYD3. Amino Acid Squence: LARKDTPKNRHPGWSR | |
0.1 mg | |
Primary | |
SPRYD3 antibody is predicted to not cross-react with other SPRYD protein family members. At least two isoforms of SPRYD3 are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction