Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRYD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558784 Supplier Invitrogen™ Supplier No. PA558784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LARKDYPKNR HPGWSRGSVA YHADDGKIFH GSGVGDPFGP RCYKGDIMGC GIMFPRDYIL DSEGDSDDSC DTVILSPTAR AVRNVRNVMY LH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The SPRY domain-containing protein 3 (SPRYD3) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. Little is known of the function of the SPRYD3 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRYD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPRYD3
Gene Accession No. Q8NCJ5
Gene Alias BC008150; RGD1561500; SPRY domain containing 3; SPRY domain-containing protein 3; SPRYD3
Gene Symbols SPRYD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPRYD3. Recombinant protein control fragment (Product #RP-97816).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223918, 84926
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.