Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPRYD5 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305859H
Description
SPRYD5 Polyclonal specifically detects SPRYD5 in Human samples. It is validated for Western Blot.Specifications
SPRYD5 | |
Polyclonal | |
Western Blot | |
MGC10977, SPRY domain containing 5, SPRY domain-containing protein 5, TRIM51, tripartite motif-containing 51, Tripartite motif-containing protein 51 | |
Produced in rabbits immunized with E. coli-derived Human SPRYD5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
84767 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction