Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRYD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585044 Supplier Invitrogen™ Supplier No. PA585044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQHHLERLRK EGEDIFQQLN ESKARMEHSR ELL Highest antigen sequence indentity to the following orthologs: Rat - 47%, Mouse - 47%.

The SPRY domain-containing protein 5 (SPRYD5) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. SPRYD5 belongs to the TRIM/RBCC family and contains one B box-type zinc finger, one B30.2/SPRY domain and one RING-type zinc finger. Little is known of the function of the SPRYD5 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRYD5
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM51
Gene Accession No. Q9BSJ1
Gene Alias SPRY domain containing 5; SPRY domain-containing protein 5; SPRYD5; TRIM51; TRIM51A; tripartite motif-containing 51; tripartite motif-containing protein 51
Gene Symbols TRIM51
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM51. Recombinant protein control fragment (Product #RP-108693).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.