Learn More
Invitrogen™ SPRYD7 Monoclonal Antibody (2G4)

Description
Peptide Sequence: MHSLVMRNDG ALYHNNEEKN RLPANSLPQE GDVVGITYDH VELNVYLNGK NMHCPASGIR GTVYPVVYVD DSAILDCQFS EFYHTPPPGF EKILFEQQIF.
Specifications
Specifications
| Antigen | SPRYD7 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2G4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Spryd7 |
| Gene Accession No. | Q5M7T2, Q5W111 |
| Gene Alias | 6330409N04Rik; C13orf1; Chronic lymphocytic leukemia deletion region gene 6 protein; chronic lymphocytic leukemia deletion region gene 6 protein homolog; CLL deletion region gene 6 protein; CLL deletion region gene 6 protein homolog; Clld6; PUR1; RGD1306437; SPRY domain containing 7; SPRY domain-containing protein 7; SPRYD7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.