Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPSB1 Monoclonal Antibody (3E7)
GREENER_CHOICE

Catalog No. PIMA522635
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522635 Supplier Invitrogen™ Supplier No. MA522635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MGQKVTGGIK TVDMRDPTYR PLKQELQGLD YCKPTRLDLL LDMPPVSYDV QLLHSWNNND RSLNVFVKED DKLIFHRHPV AQSTDAIRGK VGYTRGLHVW QITWAMRQRG THAVVGVATA DAPLHSVGYT TLVGNNHESW GWDLGRNRLY HDDKNQPSKT YPAFLEPDET FIVPDSFLVA LDMDDGTLSF IVDGQYMGVA FRGLKGKKLY PVVSAVWGHC EIRMRYLNGL DPEPLPLMDL CRRSVRLALG RERLGEIHTL PLPASLKAYL LYQ.

Substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:15601820, PubMed:21199876). Negatively regulates nitric oxide (NO) production and limits cellular toxicity in activated macrophages by mediating the ubiquitination and proteasomal degradation of NOS2 (PubMed:21199876). Acts as a bridge which links NOS2 with the ECS E3 ubiquitin ligase complex components ELOC and CUL5 (PubMed:21199876). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPSB1
Applications ELISA
Classification Monoclonal
Clone 3E7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SPSB1
Gene Accession No. Q96BD6
Gene Alias 1110014L01Rik; 4930422J18Rik; AI596360; AI854583; CBG; fi35c01; hypothetical protein LOC334190; novel SPRY domain containing protein; RGD1309319; splA/ryanodine receptor domain and SOCS box containing 1; SPRY domain-containing SOCS box protein 1; SPRY domain-containing SOCS box protein SSB-1; Spsb1; SSB1; SSB-1; Unknown (protein for IMAGE:8228399); wu:fi35c01; zgc:114115
Gene Symbols SPSB1
Host Species Mouse
Immunogen SPSB1 (AAH15711, 1 a.a. ~ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80176
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.