Learn More
Invitrogen™ SPSB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110426
Description
Immunogen sequence: MGQKVTGGIK TVDMRDPTYR PLKQELQGLD YCKPTRLDLL LDMPPVSYDV QLLHSWNNND RSLNVFVKED DKLIFHRHPV AQSTDAIRGK VGYTRGLHVW QITWAMRQRG THAVVGVATA DAPLHSVGYT TLVGNNHESW GWDLGRNRLY HDGKNQPSKT YPAFLEPDET FIVPDSFLVA LDMDDGTLSF IVDGQYMGVA FRGLKGKKLY PVVSAVWGHC EIRMRYLNGL DPEPLPLMDL CRRSVRLALG RERLGEIHTL PLPASLKAYL LYQ.
Substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:15601820, PubMed:21199876). Negatively regulates nitric oxide (NO) production and limits cellular toxicity in activated macrophages by mediating the ubiquitination and proteasomal degradation of NOS2 (PubMed:21199876). Acts as a bridge which links NOS2 with the ECS E3 ubiquitin ligase complex components ELOC and CUL5 (PubMed:21199876). [UniProt].
Specifications
| SPSB1 | |
| Polyclonal | |
| Unconjugated | |
| SPSB1 | |
| 1110014L01Rik; 4930422J18Rik; AI596360; AI854583; CBG; fi35c01; hypothetical protein LOC334190; novel SPRY domain containing protein; RGD1309319; splA/ryanodine receptor domain and SOCS box containing 1; SPRY domain-containing SOCS box protein 1; SPRY domain-containing SOCS box protein SSB-1; Spsb1; SSB1; SSB-1; Unknown (protein for IMAGE:8228399); wu:fi35c01; zgc:114115 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 74646 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D5L7 | |
| SPSB1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human SPSB1 (Q96BD6). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.