Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPSB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563089 Supplier Invitrogen™ Supplier No. PA563089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGQTALAGGS SSTPTPQALY PDLSCPEGLE ELLSAPPPDL GAQRRHGWNP KDCSENIEVK EGGLYFERR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

This gene encodes a member of a subfamily of proteins containing a central SPRY (repeats in splA and RyR) domain and a C-terminal suppressor of cytokine signaling (SOCS) box. This protein plays a role in cell signaling. This gene is present in a gene-rich cluster on chromosome 12p13 in the vicinity of the CD4 antigen and triosephosphate isomerase genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPSB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPSB2
Gene Accession No. Q99619
Gene Alias AI461677; C9; gene rich cluster, C9 gene protein; gene-rich cluster protein C9; Grcc9; splA/ryanodine receptor domain and SOCS box containing 2; SPRY domain-containing SOCS box protein 2; SPRY domain-containing SOCS box protein SSB-2; Spsb2; SSB2; SSB-2
Gene Symbols SPSB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPSB2. Recombinant protein control fragment (Product #RP-100060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.