Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPSB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561257
Description
Immunogen sequence: KVSFHMEYSC GTAAIRGTKE LGEGQHFWEI KMTSPVYGTD MMVGIGTSDV DLDKYRHTFC SLLGRDEDSW GLSYTGLLHH KGDKTSFSSR FGQGSIIGVH LDTWHGTLTF FKNRKCIGVA ATKLQNKRFY PMVC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
May be a substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
SPSB3 | |
Polyclonal | |
Unconjugated | |
SPSB3 | |
2310012N15Rik; 3300001M01Rik; C16orf31; Kiaa4204; mKIAA4204; splA/ryanodine receptor domain and SOCS box containing 3; SPRY domain-containing SOCS box protein 3; SPRY domain-containing SOCS box protein SSB-3; SPSB3; SSB3; SSB-3; Tce1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PJ21 | |
SPSB3 | |
Recombinant protein corresponding to Human SPSB3. Recombinant protein control fragment (Product #RP-102071). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction