Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPSB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584566 Supplier Invitrogen™ Supplier No. PA584566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKLSGSLKSV EVREPALRPA KRELRGAEPG RPARLDQLLD MPAAGLAVQL RH Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:21199876, PubMed:15601820). Negatively regulates nitric oxide (NO) production and limits cellular toxicity in activated macrophages by mediating the ubiquitination and proteasomal degradation of NOS2 (PubMed:21199876). Acts as a bridge which links NOS2 with the ECS E3 ubiquitin ligase complex components ELOC and CUL5 (PubMed:21199876). Diminishes EphB2-dependent cell repulsive responses by mediating the ubiquitination and degradation of EphB2/CTF2 (PubMed:28931592). Regulates cellular clock function by mediating the ubiquitin/proteasome-dependent degradation of the circadian transcriptional repressor NR1D1 (PubMed:26392558). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPSB4
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPSB4
Gene Accession No. Q96A44
Gene Alias splA/ryanodine receptor domain and SOCS box containing 4; SPRY domain-containing SOCS box protein 4; SPRY domain-containing SOCS box protein SSB-4; SPSB4; SSB4; SSB-4
Gene Symbols SPSB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPSB4. Recombinant protein control fragment (Product #RP-107902).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92369
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.