Learn More
Spt16 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302492AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT , interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.Specifications
Spt16 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y5B9 | |
SUPT16H | |
Between 997 and 1047 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SUPT16H | |
SUPT16H; FACT; suppressor of Ty 16 homolog (S. cerevisiae); HGNC:11465; CDC68; FACTP140; FLJ10857; FLJ14010; SPT16/CDC68; chromatin-specific transcription elongation factor large subunit; FLJ34357; facilitates chromatin remodeling 140 kDa subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11198 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.