Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPT3 Monoclonal Antibody (OTI3D1)
GREENER_CHOICE

Catalog No. PIMA526938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526938 Supplier Invitrogen™ Supplier No. MA526938
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPT3 Monoclonal Antibody for Western Blot, IHC (P)

The transcription of many RNA polymerase II-dependent genes requires Spt3, a member of the S. cerevisiae SAGA complex. Transcription from delta sequences, the long terminal repeats that flank yeast Ty elements, requires the yeast SPT3 gene. Spt3 and Spt20 work together to recruit TATA-box binding protein (TBP) to the core promoter allowing TBP to bind to SAGA-dependent promoters. Null mutations in the Spt3 gene cause defects in sporulation, diploid filamentous growth, and haploid invasive growth, indicating that Spt3 has an important role in both mating and development pathways in yeast. At the promoters of some genes including yeast HO, HIS3 and TRP3 genes, Spt3 inhibits binding of TBP, resulting in reduced transcription. This repressive effect of Spt3 can be overcome by another member of the SAGA complex, GCN5, which promotes the formation of a TBP/TFIIA complex by histone acetylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPT3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SUPT3H
Gene Accession No. O75486
Gene Alias Cdc5-like protein; D9509.12; Positive regulator of Ty transcription; Protein SPT3; SPT3; SPT3 homolog, SAGA and STAGA complex component; SPT3L; SPT3-like protein; suppressor of Ty 3 homolog; suppressor of Ty 3 homolog (S. cerevisiae); SUPT3H; Transcription initiation protein SPT3 homolog; transcriptional regulator SPT3; YDR392W
Gene Symbols SUPT3H
Host Species Mouse
Immunogen Full length human recombinant protein of SUPT3H produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8464
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.