Learn More
Invitrogen™ SPT5 Polyclonal Antibody

Description
Immunogen sequence: VGYSPMTPGA PSPGGYNPHT PGSGIEQNSS DWVTTDIQVK VRDTYLDTQV VGQTGVIRSV TGGMCSVYLK DSEKVVSISS EHLEPITPTK NNKVKVILGE DREATGVLLS IDGEDGIVRM DLDEQLKILN LRFL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
Specifications
| Antigen | SPT5 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SUPT5H |
| Gene Accession No. | O00267, O55201 |
| Gene Alias | AL033283; AL033321; AU019549; DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; etID10129.15; etID37101.15; fb37g01; fb95e05; fog; foggy; hSPT5; protein foggy; Spt5; SPT5 homolog, DSIF elongation factor subunit; SPT5H; suppressor of Ty 5; suppressor of Ty 5 homolog; suppressor of Ty 5 homolog (S. cerevisiae); Supt5; Supt5h; supt5h protein; Tat-cotransactivator 1 protein; Tat-CT1; Tat-CT1 protein; Transcription elongation factor SPT5; transcription elongation regulator FOGGY; wu:fb37g01; wu:fb95e05; wu:fc10f02 |
| Gene Symbols | Supt5, SUPT5H |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.