Learn More
SPTAN1/Alpha II-spectrin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
| Antigen | SPTAN1/Alpha II-spectrin |
|---|---|
| Applications | Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1776
|
Bethyl Laboratories, Inc
IHC00259T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1775
|
Bethyl Laboratories, Inc
IHC00259 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, X. tropicalis, Bovine, Dog, Horse, Guinea pig_10141, Pig, Panda, Or
Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. This gene encodes an alpha spectrin that is specifically expressed in nonerythrocytic cells. The encoded protein has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.Specifications
| SPTAN1/Alpha II-spectrin | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| (ALPHA)II-SPECTRIN, FLJ44613, NEAS | |
| SPTAN1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13813 | |
| 6709 | |
| Between 2422 and 2472 | |
| Primary | |
| 4° C | |
| SPTAN1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.