Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPTAN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$514.50 - $1763.00
Specifications
Antigen | SPTAN1 |
---|---|
Concentration | Conc. Not Determined |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143496
|
Invitrogen™
PA5143496 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143594
|
Invitrogen™
PA5143594 |
100 μL |
Each for $514.50
|
|
|||||
Description
SPTAN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
SPTAN1 (spectrin alpha chain, non-erythrocytic 1) is a part of the spectrin family of filamentous cytoskeletal proteins. They function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. SPTAN1 has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.Specifications
SPTAN1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat, Bovine | |
SPTAN1 | |
(ALPHA)II-SPECTRIN; 2610027H02Rik; A2a; a--fodrin; alpha; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; brain; brain alpha-spectrin; EIEE5; FLJ44613; Fodrin alpha chain; inhibitory protein factor; IPF; NEAS; noerythroid alpha-spectrin 2; Non erythrocytic spectrin alpha; non-erythrocytic 1 (alpha-fodrin); non-erythroid alpha chain; nonerythroid alpha-spectrin; nonerythroid alpha-spectrin 2; putative alpha-spectrin; SPECA; Spectrin; Spectrin alpha; spectrin alpha chain; Spectrin alpha chain brain; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; Spna2; Spna-2; Spta2; Sptan1 | |
SPTAN1 | |
Primary |
Conc. Not Determined | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
whole serum with 5mM sodium azide | |
P16086, P16546, Q13813 | |
20740, 540108, 64159, 6709 | |
Recombinant constructs spanning most of human alpha-II spectrin expressed in and purified from E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title