Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTAN1 Polyclonal Antibody

Catalog No. PIPA5143496
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143496 500 μL
PIPA5143594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143496 Supplier Invitrogen™ Supplier No. PA5143496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTAN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC

SPTAN1 (spectrin alpha chain, non-erythrocytic 1) is a part of the spectrin family of filamentous cytoskeletal proteins. They function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. SPTAN1 has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTAN1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 5mM sodium azide
Gene SPTAN1
Gene Accession No. P16086, P16546, Q13813
Gene Alias (ALPHA)II-SPECTRIN; 2610027H02Rik; A2a; a--fodrin; alpha; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; brain; brain alpha-spectrin; EIEE5; FLJ44613; Fodrin alpha chain; inhibitory protein factor; IPF; NEAS; noerythroid alpha-spectrin 2; Non erythrocytic spectrin alpha; non-erythrocytic 1 (alpha-fodrin); non-erythroid alpha chain; nonerythroid alpha-spectrin; nonerythroid alpha-spectrin 2; putative alpha-spectrin; SPECA; Spectrin; Spectrin alpha; spectrin alpha chain; Spectrin alpha chain brain; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; Spna2; Spna-2; Spta2; Sptan1
Gene Symbols SPTAN1
Host Species Rabbit
Immunogen Recombinant constructs spanning most of human alpha-II spectrin expressed in and purified from E. coli.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20740, 540108, 64159, 6709
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.