Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTBN1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA576518

Catalog No. PIPA576518


Only null left
Add to Cart

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Spectrin (Sp), the most abundant of the erythrocyte membrane skeleton proteins, helps these cells maintain their characteristic biconcave shape while remaining flexible and elastic. Erythrocyte Sp is a heterodimer composed of a 280 kDa alpha subunit and a 246 kDa beta subunit which associate in a side-to-side, antiparallel configuration to form a 100 nm rod-like structure. Sp in other tissues may be composed of distinct but homologous alpha and beta subunits, sometimes referred to as fodrin. A newly introduced nomenclature designates the Sp subunits of the erythrocyte as alpha-1 and beta-1, and the fodrin subunits as alpha-2 and beta-2. Alternatively spliced forms of each are designated as epsilon-1, epsilon-2, etc. (e. g. beta-1 epsilon-1).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPTBN1
Polyclonal
Unconjugated
SPTBN1
9930031C03Rik; A2a; a--fodrin; AI842465; AL033301; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin 2; beta fodrin; beta II spectrin-like (short form); beta-fodrin; beta-G spectrin; beta-I spectrin; Beta-II spectrin; beta-spectrin 1; beta-spectrin 2; beta-spectrin 2, non-erythrocytic; beta-spectrin II; beta-spectrin non-erythrocytic 1; betaSpII; brain alpha-spectrin; brain erythroid spectrin (235E); brain spectrin; bSPII-; cytoskeleton protein; D330027P03Rik; EL3; ELF; elf1; elf3; embryonic liver beta-fodrin; embryonic liver fodrin; embryonic liver fodrin beta chain; epididymis luminal protein 102; erythroid spectrin beta; fodrin alpha chain; Fodrin beta chain; Gm1301; HEL102; HS2; HSPTB1; inhibitory protein factor; IPF; ja; jaundiced; membrane cytoskeletal protein; mKIAA4049; mKIAA4219; noerythroid alpha-spectrin 2; non-erythrocytic; nonerythroid alpha-spectrin 2; non-erythroid spectrin beta; short form of beta II spectrin; Sp beta; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin beta 1; spectrin beta 2; spectrin beta chain, brain 1; spectrin beta chain, erythrocyte; spectrin beta chain, erythrocytic; Spectrin beta chain, non-erythrocytic 1; spectrin beta Tandil; spectrin beta, erythrocytic; spectrin beta, non-erythrocytic 1; spectrin G; spectrin R; spectrin, alpha, non-erythrocytic 1; spectrin, beta, erythrocytic; spectrin, beta, non-erythrocytic 1; spectrin, non-erythroid alpha chain; spectrin, non-erythroid beta chain 1; SPH2; Spna2; Spnb1; Spnb-1; Spnb2; Spnb-2; Spta2; Sptan1; SPTB; SPTB1; Sptb2; Sptbn1
Rabbit
Antigen affinity chromatography
RUO
20742, 6711
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q01082, Q62261
SPTBN1
Recombinant fusion protein of human SPTBN1.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.