Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPTB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582088
Description
Immunogen sequence: TSVLILQRKH KAFEDELRGL DAHLEQIFQE AHGMVARKQF GHPQIEARIK EVSAQWDQLK DLAAFCKKNL QDAENFFQFQ GDADDLKAWL QDAHRLLSGE DVGQDEGATR ALGKKHKDFL EELEESRGVM EHLEQQAQGF PEEF.
SPTB (Spectrin Beta, Erythrocytic) is a Protein Coding gene. Diseases associated with SPTB include Spherocytosis, Type 2 and Elliptocytosis 3. Among its related pathways are Cytokine Signaling in Immune system and RET signaling. Gene Ontology (GO) annotations related to this gene include protein heterodimerization activity and actin filament binding. An important paralog of this gene is SPTBN1.
Specifications
SPTB | |
Polyclonal | |
Unconjugated | |
SPTB | |
HSPTB1; Spectrin Beta Erythrocytic; Spectrin Beta, Erythrocytic; SPTB1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11277 | |
SPTB | |
Recombinant protein corresponding to Human SPTB. Recombinant protein control fragment (Product #RP-89116). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction