Learn More
SPTBN1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00157

Description
The recommended shelf life for this product is 1 year from date of receipt.
Spectrin is an actin crosslinking and molecular scaffold protein that links the plasma membrane to the actin cytoskeleton, and functions in the determination of cell shape, arrangement of transmembrane proteins, and organization of organelles. It is composed of two antiparallel dimers of alpha- and beta- subunits. This gene is one member of a family of beta-spectrin genes. The encoded protein contains an N-terminal actin-binding domain, and 17 spectrin repeats which are involved in dimer formation. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SPTBN1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q01082 | |
SPTBN1 | |
Between 2150 and 2200 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
SPTBN1 | |
SPTB2, betaSpII, spectrin, beta, non-erythrocytic 1, ELF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6711 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.