Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPTBN2 Mouse anti-Human, Clone: SPTBN2/1582, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14993
Description
Mouse monoclonal antibody raised against partial recombinant human SPTBN2.
- High specificity
- High reactivity
Specifications
SPTBN2 | |
Monoclonal | |
Unconjugated | |
ELISA (2-4 ug/mL for coating) Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal work | |
O15020 | |
SPTBN2 | |
Recombinant protein corresponding to amino acids 356-475 of human SPTBN2. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
SPTBN2/1582 | |
Mouse monoclonal antibody raised against partial recombinant human SPTBN2. | |
In 10 mM PBS. | |
GTRAP41/SCA5 | |
Mouse | |
Protein A/G purification | |
RUO | |
6712 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction