Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN2 Rabbit anti-Human, Unconjugated, Clone: SPTBN2/2887R, Abnova™

Catalog No. 89339612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-612 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-612 Supplier Abnova Corporation Supplier No. RAB00633
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SPTBN2 Monoclonal antibody specifically detects SPTBN2 in Human samples. It is validated for WB-Ce, IF, Flow Cyt.

Specifications

Antigen SPTBN2
Applications Western Blot, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone SPTBN2/2887R
Conjugate Unconjugated
Description spectrin, beta, non-erythrocytic 2
Gene SPTBN2
Gene Alias GTRAP41, SCA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 356-475 of human SPTBN2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.