Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPTBN4, Mouse anti-Human, Biotin, Clone: S393-2, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16461
Specifications
SPTBN4 | |
Monoclonal | |
Biotin | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
KIAA1642/QV/SPNB4/SPTBN3 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
S393-2 | |
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
spectrin, beta, non-erythrocytic 4 | |
SPTBN4 | |
Recombinant protein corresponding to amino acids 1621-1832 at C-terminus of human SPTBN4. | |
RUO | |
57731 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction