Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN4 Mouse anti-Human, Mouse, Rat, ATTO 390, Clone: S393-2, Abnova™

Catalog No. 89338936 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-936 Supplier Abnova Corporation Supplier No. MAB19432
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

SPTBN4 Monoclonal antibody specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for WB, ICC, IHC-P, IF.

Specifications

Antigen SPTBN4
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S393-2
Conjugate Atto 390
Description spectrin, beta, non-erythrocytic 4
Gene SPTBN4
Gene Alias KIAA1642, QV, SPNB4, SPTBN3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1621-1832 at C-terminus repeats 14 to 15 of human Beta4-spectrin
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 57731
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.