Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTLC1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017239 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017239 Supplier Novus Biologicals Supplier No. NBP281928
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTLC1 Polyclonal specifically detects SPTLC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPTLC1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.3.1.50, hereditary sensory neuropathy, type 1, HSAN1, LBC1, LCB 1, LCB1HSAN, Long chain base biosynthesis protein 1, MGC14645, serine C-palmitoyltransferase, serine palmitoyltransferase 1, serine palmitoyltransferase, long chain base subunit 1, Serine-palmitoyl-CoA transferase 1, SPT 1, SPT1HSN1, SPTI
Gene Symbols SPTLC1
Host Species Rabbit
Immunogen SPTLC1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SPTLC1. The immunogen is located within amino acids 380 - 430 of SPTLC1. Amino Acid Squence: HLQLEESTGSREQDVRL
Molecular Weight of Antigen 51 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10558
Test Specificity At least two isoforms of SPTLC1 are known to exist; this antibody will detect only isoform a. SPTLC1 antibody is predicted to not cross-react with SPT2.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.