Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTLC1 Mouse anti-Human, Unconjugated, Clone: 2D11D7, Proteintech
SDP

Catalog No. 502239087 Shop All Proteintech Group Inc Products
Encompass
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9087 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9087 Supplier Proteintech Group Inc Supplier No. 668991IG150UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The SPTLC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SPTLC1. This antibody recognizes human, mouse, rat antigen. The SPTLC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen SPTLC1
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 2D11D7
Concentration 500 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 10558
Target Species Human, Rat, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.