Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542914 Supplier Invitrogen™ Supplier No. PA542914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ICPLPELVKL KYKYKARIFL EESLSFGVLG EHGRGVTEHY GINIDDIDLI Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 93%.

Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Sptlc1
Gene Accession No. O15269
Gene Alias AW552086; C77762; E030036H05; fc75h04; fp41c08; hLCB1; HSAN; HSAN1; HSN1; hypothetical protein LOC553981; hypothetical protein LOC614165; LBC1; LCB 1; LCB1; long chain base biosynthesis protein 1; MGC14645; rCG_44191; RGD1306617; RP11-118F2.1; serine C-palmitoyltransferase; serine palmitoyltransferase 1; serine palmitoyltransferase long chain base subunit 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; serine-palmitoyl-CoA transferase 1; SPT 1; Spt1; SPTI; sptlc1; wu:fc75h04; wu:fp41c08; zgc:112247
Gene Symbols Sptlc1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SPTLC1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10558
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.