Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPTLC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551091
Description
SPTLC1 Polyclonal Antibody detects endogenous levels of SPTLC1 protein.
Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
Specifications
SPTLC1 | |
Polyclonal | |
Unconjugated | |
Sptlc1 | |
AW552086; C77762; E030036H05; fc75h04; fp41c08; hLCB1; HSAN; HSAN1; HSN1; hypothetical protein LOC553981; hypothetical protein LOC614165; LBC1; LCB 1; LCB1; long chain base biosynthesis protein 1; MGC14645; rCG_44191; RGD1306617; RP11-118F2.1; serine C-palmitoyltransferase; serine palmitoyltransferase 1; serine palmitoyltransferase long chain base subunit 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; serine-palmitoyl-CoA transferase 1; SPT 1; Spt1; SPTI; sptlc1; wu:fc75h04; wu:fp41c08; zgc:112247 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10558, 268656, 361213 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15269, O35704 | |
Sptlc1 | |
Synthesized peptide derived from the C-terminal region of human SPTLC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction