Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121032 Supplier Invitrogen™ Supplier No. PA5121032
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SKOV3, MCF7, HepG2, Mouse kidney, Mouse lung, Mouse liver, Mouse thymus, Rat lung, Rat kidney. The target is usually found in the following locations: Endoplasmic reticulum membrane, Single-pass membrane protein. Immunogen sequence: RLLFSKTYKL QERSDLTVKE KEELIEEWQP EPLVPPVPKD HPALNYNIVS GPPSHKTVVN GKECINFASF NFLGLLDNPR VKAAALASLK KYGVGTCGPR GFYGTFDVHL DLEDRLAKFM KTEEAIIYSY GFATIASAIP AYSKRGDIVF VDRAACFAIQ KGLQASRSDI KLFKHNDMAD LERLLKEQEI EDQKNPRKAR VTRRFIVVEG LYMNTGTICP LPELVKLKYK YKARIFLEES LSFGVLGEHG RGVTEHYGIN IDDIDLISAN MENA.

Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sptlc1
Gene Accession No. O15269, O35704
Gene Alias AW552086; C77762; E030036H05; fc75h04; fp41c08; hLCB1; HSAN; HSAN1; HSN1; hypothetical protein LOC553981; hypothetical protein LOC614165; LBC1; LCB 1; LCB1; long chain base biosynthesis protein 1; MGC14645; rCG_44191; RGD1306617; RP11-118F2.1; serine C-palmitoyltransferase; serine palmitoyltransferase 1; serine palmitoyltransferase long chain base subunit 1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; serine-palmitoyl-CoA transferase 1; SPT 1; Spt1; SPTI; sptlc1; wu:fc75h04; wu:fp41c08; zgc:112247
Gene Symbols Sptlc1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-310 of human SPTLC1 (NP_006406.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10558, 268656, 361213
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.