Learn More
SPTLC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | SPTLC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5050
![]() |
Bethyl Laboratories, Inc
A303407AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Serine palmitoyltransferase, which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It converts L-serine and palmitoyl-CoA to 3-oxosphinganine with pyridoxal 5'-phosphate as a cofactor. SPT1 is the long chain base subunit 1 of mammalian serine palmitoyltransferase. SPT1 is not catalytically active but is necessary for the stabilization of the SPT2 subunit and anchoring the holoenzyme to the cytosolic face of the endoplasmic reticulum. Missense mutations in this gene have been identified in patients with hereditary sensory neuropathy type 1 (HSAN1). These mutations induce a shift in the substrate specificity of the holoenzyme, leading to the formation and accumulation of two neurotoxic sphingolipids.Specifications
SPTLC1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Serine palmitoyltransferase 1; Long chain base biosynthesis protein 1; LCB 1; Serine-palmitoyl-CoA transferase 1; SPT 1; SPT1 | |
Between 50 and 100 | |
Primary | |
4° C | |
SPTLC1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O15269, O35704 | |
10558, 268656 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.