Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTLC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531130 Supplier Invitrogen™ Supplier No. PA531130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse liver. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (80%), Chicken (84%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Serine palmitoyltransferase (SPT), which consists of two different subunits, is the key enzyme in sphingolipid biosynthesis. It converts L-serine and palmitoyl-CoA to 3-oxosphinganine with pyridoxal 5'-phosphate as a cofactor. SPT2 is the long chain base subunit 2 of mammalian serine palmitoyltransferase. SPT2 is catalytically active but needs its related protein SPT1 for its stabilization and anchoring the holoenzyme to the cytosolic face of the endoplasmic reticulum. As in the case with SPT1, mutations in the SPT2 gene can cause hereditary sensory and autonomic neuropathy type I (HSAN-I), resulting from a loss of SPT activity as well as the accumulation of the atypical and neurotoxic sphingoid metabolite 1-deoxy-sphinganine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SPTLC2
Gene Accession No. O15270, P97363
Gene Alias AI173915; hLCB2a; HSN1C; KIAA0526; LCB 2; LCB2; LCB2a; long chain base biosynthesis protein 2; Long chain base biosynthesis protein 2a; mKIAA0526; NSAN1C; Pomt2; protein-O-mannosyltransferase 2; serine palmitoyltransferase 2; serine palmitoyltransferase long chain base subunit 2; serine palmitoyltransferase, long chain base subunit 2; serine palmitoyltransferase, subunit II; serine-palmitoyl-CoA transferase 2; SPT 2; SPT2; Sptlc2
Gene Symbols SPTLC2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Serine palmitoyltransferase 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20773, 9517
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.