Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SPTLC2, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1641MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1641MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1641MI Supplier MilliporeSigma™ Supplier No. ABS1641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serine palmitoyltransferase 2 (EC 2.3.1.50; UniProt O15270; also known as hLCB2a, LCB 2, LCB2a, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, Serine-palmitoyl-CoA transferase 2, SPT 2) is encoded by the SPTLC2 (also known as HSAN1C, KIAA0526, NSAN1C, LCB2) gene (Gene ID 9517) in human. Serine palmitoyltransferase (SPT) is a pyridoxal 5&prime-phosphate (PLP)-dependent enzyme that catalyses the first step of de novo sphingolipids (SLs) biosynthesis, namely the condensation of L-serine with palmitoyl coenzyme-A to generate 3-ketodehydrosphinganine (KDS) that forms the sphingoid base backbone of all SLs. The core human SPT homoenzyme is a membrane-bound heterodimer composed of an hLCB1 (SPTLC1) subunit together with either an hLCB2a (SPTLC2) or an hLCB2b (SPTLC3) subunit. The hLCB1/hLCB2 heterodimer contains a single active site composed of residues from both subunits. hLCB2a and hLCB2b share 68% sequence identity, with hLCB2b being the isotype that is predominantly expressed in the placenta and prefers shorter chain acyl-CoAs (C12 and C14) to generate short-chain SLs. In addition, two human small subunits (ssSPTa and ssSPTb) have been identified and found to maximize the catalytic activity and to confer the acyl-CoA substrate chain-length specificity of the hLCB1/hLCB2 core enzyme. Mutations in hLCB1 and hLCB2a are found in patients with hereditary sensory and autonomic neuropathy type I (HSAN1), an inherited disorder that affects sensory and autonomic neurons due to neurotoxic deoxysphingolipids formation as a result of substrate promiscuity of the mutated SPT enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene SPTLC2, HSAN1C, KIAA0526, NSAN1C, LCB2
Gene Accession No. O15270
Host Species Rabbit
Immunogen A mixture of two linear peptides corresponding to sequences from the N-terminal region and the C-terminal end of human SPTLC2.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004854
Test Specificity Target-specificity of this antiserum was verified by negative blotting using pre-immune serum and by target band blocking using immunogen peptides (Hornemann, T., et al. (2006). J. Biol. Chem. 281(49):37275-37281).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.