Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTLC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598858
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598858 Supplier Invitrogen™ Supplier No. PA598858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTLC3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Serine palmitoyltransferase (SPT). The heterodimer formed with LCB1/SPTLC1 constitutes the catalytic core. The composition of the serine palmitoyltransferase (SPT) complex determines the substrate preference. The SPTLC1-SPTLC3-SPTSSA isozyme uses both C14-CoA and C16-CoA as substrates, while the SPTLC1-SPTLC3-SPTSSB has the ability to use a broader range of acyl-CoAs without apparent preference.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTLC3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPTLC3
Gene Accession No. Q8BG54, Q9NUV7
Gene Alias C130053K05Rik; C20orf38; dJ718P11; dJ718P11.1; hLCB2b; LCB 3; LCB2b; LCB3; long chain base biosynthesis protein 2b; Long chain base biosynthesis protein 3; serine palmitoyltransferase 3; serine palmitoyltransferase long chain base subunit 3; serine palmitoyltransferase, long chain base subunit 2-like (aminotransferase 2); serine palmitoyltransferase, long chain base subunit 3; serine-palmitoyl-CoA transferase 3; Serine-palmityl-CoA transferase 3; SPT 3; SPT3; SPTLC2L; SPTLC3
Gene Symbols SPTLC3
Host Species Rabbit
Immunogen Recombinant Human Serine palmitoyltransferase 3 protein (1-58AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228677, 55304
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.