Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPTY2D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565486
Description
Immunogen sequence: SGPGRSISGSI PAGRTVSNSV PGRPVSSLGP GQTVSSSGPT IKPKCTVVSE TISSKNIISR SSNGQMNGMK PPLSGYRAAQ GPQRLPFPTG YKRQREYEE Highest antigen sequence identity to the following orthologs - mouse 86%, rat 83%.
Histone chaperone that stabilizes pre-existing histone tetramers and regulates replication-independent histone exchange on chromatin (PubMed:26109053). Required for normal chromatin refolding in the coding region of transcribed genes, and for the suppression of spurious transcription (PubMed:26109053). Binds DNA and histones and promotes nucleosome assembly (in vitro) (PubMed:23378026, PubMed:26109053). Facilitates formation of tetrameric histone complexes containing histone H3 and H4 (PubMed:26109053). Modulates RNA polymerase 1-mediated transcription. Binds DNA, with a preference for branched DNA species, such as Y-form DNA and Holliday junction DNA (PubMed:23378026).
Specifications
SPTY2D1 | |
Polyclonal | |
Unconjugated | |
SPTY2D1 | |
DKFZp686I068; FLJ39441; Protein KU002155; Protein SPT2 homolog; Spt2; SPT2 chromatin protein domain containing 1; SPT2 domain-containing protein 1; SPT2, Suppressor of Ty, domain containing 1; SPTY2D1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
144108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68D10 | |
SPTY2D1 | |
Recombinant Human SPTY2D1. Recombinant protein control fragment (Product #RP-104840). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction